Learn More
Honeywell™ Whiting & Davis Metal Mesh Gloves
Catalog No. p-141668
Click to view available options
Size:
X-Small
Small
Medium
Large
This item is not returnable. View return policy
Catalog No. 19-999-885
Supplier: Honeywell™ 5902XS E
This item is not returnable. View return policy
Made of stainless-steel ring mesh; Highest level of cut protection; Reversible glove with rubber band straps for a snug fit.
Entire glove is made of a mesh of high tensile-strength, corrosion-resistant stainless-steel rings, each individually welded for maximum strength and remarkable flexibility. Provides the highest level of protection against cuts and lacerations in cutting operations. Ergonomic design offers optimal comfort and fit; fingers are tapered and glove body is contoured to the shape of the hand. Self-adjusting elastic band grips wrist snugly, holding glove in place. Reversibility maximizes use and reduces inventory costs. Stainless-steel rings resist process oils and fats, and are easy to clean.
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size X-Small
For Use With (Application) Meat, poultry, fish processing, food service, knife and blade sharpening/cleaning, fishing, garment manufacturing, veterinary services, animal control, paper and pulp industry
Glove Cuff Style Elasticated
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.