Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ WorkEasy™ 50 Lite Polyurethane Coated Nylon Gloves

Catalog No. 19160007
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Lightweight nylon gloves provide protection against abrasion, while polyurethane coating on the palm/fingers ensures tactile sensitivity.

  • 18 gauge lightweight liner and coating ensures tactile sensitivity
  • Polyurethane coating helps to reduce abrasion, snags, and tears in the glove
  • Launderable for extended useful life
TRUSTED_SUSTAINABILITY

Specifications

Construction Seamless Knit
Coating Material Polyurethane
Color Gray, Blue
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Small
For Use With (Application) Small Parts Handling, Electronics Assembly, and Painting
Glove Cuff Style Knit
Series WE50
Yarn Gauge 18
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.