Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ WorkEasy™ 300 Dyneema™ Cut-Resistant Gloves, Polyurethane Coated
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Description
- High level of cut protection for hazardous applications
- Lightweight fabric blend provides the same level of protection, without the bulk to prevent hand fatigue
- Polyurethane coating provides abrasion resistance and enhanced grip
- Launderable for extended glove life

Specifications
Specifications
Coating Material | Polyurethane |
Color | Gray |
Cut Resistance Rating | ANSI A3 |
Exterior Finish | No Texture |
Hand | Specific |
Material | Dyneema Fiber |
Product Type | Cut-resistant Gloves |
Size | 2X-Large |
For Use With (Application) | Small parts assembly, fabrication, inspection, warehousing |
Glove Cuff Color | White |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction