Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Hook Collar Clamp
SDPSCI_ED

Catalog No. p-3075064
Click to view available options
For Use With (Equipment):
1/2 in. diameter rods
3/4 in. diameter rods

Hook Collar Clamp for suspending pulleys, balances, etc. from support rods or cross members.

  • Plated steel collar clamps
  • Thumbscrews and hooks for suspending pulleys, balances, etc. from support rods or crossbars.
  • For 1/2 in. to 3/4 in. diameter rods

Specifications

Type Hose and Tubing Clamp
Material Steel
For Use With (Equipment) 1/2 in. diameter rods
For Use With (Application) For suspending pulleys, balances etc. from support rods or cross members 0.5 in.
Description Hook Collar Clamp, for 1/2 in. diameter rods
Color Silver
Jaw Type Fixed
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.