Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOOK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553836
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553836 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553836 Supplier Invitrogen™ Supplier No. PA553836
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KRALEELQEA LAEKEELRQR CEELDMQVTT LQDEKNSLVS ENEMMNEKLD QLDGSFDDPN TVVAKKYFHA QLQLEQLQEE NFRLEAAKDD YRVHC Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

This gene encodes a member of the hook family of coiled-coil proteins, which bind to microtubules and organelles through their N- and C-terminal domains, respectively. The encoded protein localizes to discrete punctuate subcellular structures, and interacts with several members of the Rab GTPase family involved in endocytosis. It is thought to link endocytic membrane trafficking to the microtubule cytoskeleton. Several alternatively spliced transcript variants have been identified, but the full-length nature of some of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOOK1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HOOK1
Gene Accession No. Q9UJC3
Gene Alias A930033L17Rik; abnormal spermatozoon head shape; azh; hHK1; h-hook1; HK1; hook homolog 1; hook homolog 1 (Drosophila); hook microtubule-tethering protein 1; HOOK1; mHK1; protein Hook homolog 1
Gene Symbols HOOK1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HOOK1. Recombinant protein control fragment (Product #RP-92295).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51361
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.