Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOOK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599038
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599038 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599038 Supplier Invitrogen™ Supplier No. PA599038
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HOOK1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a member of the hook family of coiled-coil proteins, which bind to microtubules and organelles through their N- and C-terminal domains, respectively. The encoded protein localizes to discrete punctuate subcellular structures, and interacts with several members of the Rab GTPase family involved in endocytosis. It is thought to link endocytic membrane trafficking to the microtubule cytoskeleton. Several alternatively spliced transcript variants have been identified, but the full-length nature of some of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOOK1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene HOOK1
Gene Accession No. Q9UJC3
Gene Alias A930033L17Rik; abnormal spermatozoon head shape; azh; hHK1; h-hook1; HK1; hook homolog 1; hook homolog 1 (Drosophila); hook microtubule-tethering protein 1; HOOK1; mHK1; protein Hook homolog 1
Gene Symbols HOOK1
Host Species Rabbit
Immunogen Recombinant Human Protein Hook homolog 1 protein (301-600AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51361
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.