Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOOK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562176
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562176 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562176 Supplier Invitrogen™ Supplier No. PA562176
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AIQELMTKDT PDSLSPETYG NFDSQSRRYY FLSEEAEEGD ELQQRCLDLE RQLMLLSEEK QSLAQENAGL RERMGRPEGE GTPGLTAKKL LLLQ Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 84%.

Hook proteins are cytosolic coiled-coil proteins that contain conserved N-terminal domains, which attach to microtubules, and more divergent C-terminal domains, which mediate binding to organelles. The Drosophila Hook protein is a component of the endocytic compartment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOOK2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HOOK2
Gene Accession No. Q96ED9
Gene Alias A630054I03Rik; hHK2; h-hook2; HK2; hook homolog 2; hook homolog 2 (Drosophila); hook microtubule tethering protein 2; hook microtubule-tethering protein 2; HOOK2; mHK2; protein Hook homolog 2
Gene Symbols HOOK2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HOOK2. Recombinant protein control fragment (Product #RP-98906).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29911
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.