Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOOK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598682
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598682 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598682 Supplier Invitrogen™ Supplier No. PA598682
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HOOK2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Hook proteins are cytosolic coiled-coil proteins that contain conserved N-terminal domains, which attach to microtubules, and more divergent C-terminal domains, which mediate binding to organelles. The Drosophila Hook protein is a component of the endocytic compartment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOOK2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HOOK2
Gene Accession No. Q7TMK6, Q96ED9
Gene Alias A630054I03Rik; hHK2; h-hook2; HK2; hook homolog 2; hook homolog 2 (Drosophila); hook microtubule tethering protein 2; hook microtubule-tethering protein 2; HOOK2; mHK2; protein Hook homolog 2
Gene Symbols HOOK2
Host Species Rabbit
Immunogen Recombinant Human Protein Hook homolog 2 protein (440-719AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170833, 29911
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.