Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOOK3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572889
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572889 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572889 Supplier Invitrogen™ Supplier No. PA572889
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HOOK3 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Hook proteins are cytosolic coiled-coil proteins that contain conserved N-terminal domains, which attach to microtubules, and more divergent C-terminal domains, which mediate binding to organelles. The Drosophila Hook protein is a component of the endocytic compartment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOOK3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene HOOK3
Gene Accession No. Q86VS8, Q8BUK6
Gene Alias 5830454D03Rik; AI317159; E330005F07Rik; hHK3; h-hook3; HK3; hook homolog 3; hook homolog 3 (Drosophila); hook microtubule-tethering protein 3; Hook3; mHK3; protein Hook homolog 3
Gene Symbols HOOK3
Host Species Rabbit
Immunogen HOOK3 antibody for a 19 amino acid peptide from near the amino terminus of human HOOK3. The immunogen is within amino acids 210 - 260 of HOOK3.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 306548, 320191, 84376
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.