Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HOOK3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598559
Description
HOOK3 Polyclonal Antibody for Western Blot, IHC (P), IP, ELISA
Hook proteins are cytosolic coiled-coil proteins that contain conserved N-terminal domains, which attach to microtubules, and more divergent C-terminal domains, which mediate binding to organelles. The Drosophila Hook protein is a component of the endocytic compartment.
Specifications
HOOK3 | |
Polyclonal | |
Unconjugated | |
HOOK3 | |
5830454D03Rik; AI317159; E330005F07Rik; hHK3; h-hook3; HK3; hook homolog 3; hook homolog 3 (Drosophila); hook microtubule-tethering protein 3; Hook3; mHK3; protein Hook homolog 3 | |
Rabbit | |
Protein G | |
RUO | |
84376 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
4.83 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q86VS8 | |
HOOK3 | |
Recombinant Human Protein Hook homolog 3 protein (357-455AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction