Learn More
HOOK3 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304906A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Hook proteins are cytosolic coiled-coil proteins that contain conserved N-terminal domains, which attach to microtubules, and more divergent C-terminal domains, which mediate binding to organelles. The Drosophila Hook protein is a component of the endocytic compartment.Specifications
HOOK3 | |
Polyclonal | |
Unconjugated | |
HOOK3 | |
hHK3, h-hook3, HK3, hook homolog 3, hook homolog 3 (Drosophila) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
320191, 84376 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q86VS8, Q8BUK6 | |
HOOK3 | |
Between 425 and 475 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.