Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Hopen™ Hygiene Face Masks

Catalog No. 19910684 Shop All Medicom Products
Click to view available options
Construction:
1-ply
2-ply
Color:
Blue
White
Material:
Paper
Polypropylene
This item is not returnable. View return policy
This item is not returnable. View return policy

General hygiene masks protects workers and environment from accidental mouth and nasal contamination.

  • Hygiene masks are economical and can be used in a variety of industries including catering or for visitors
  • Protects working environment and products manufactured by limiting the propagation of accidental mouth and nasal sprays (sneezing, coughing)
  • Comfortable due to non-woven material that repels water
  • Ensures a steady hold during extended wear times
  • Available in white paper material and blue or white polypropylene material

Specifications

Product Type General Purpose Face Mask
Color White
Fastening Type Earloops
Material Polypropylene
Sterility Non-sterile
Style Pleated
Construction 2-ply
Disposable Single-use
For Use With (Application) General hygiene in work environments, visitors
Model 60.101
Series Hopen™
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.