Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Hopen™ Pocketed, Single Use Hygiene Gown

Catalog No. 19910639
Click to view available options
Color:
Blue
White
Closure Type:
Press Studs
Zipper
Size:
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Medicom™ Hopen™ Pocketed, Single Use Hygiene Gown

  • Single use hygiene gowns protect the working environment from particles coming from the body of the operator (dander, hair, sweating ...)
  • They are suitable to HACCP procedures.
  • They allow the operator to protect himself from the risks of soiling related to the activity he carries out.

Specifications

Packaging Type Individual, Bag
No. of Pockets 2
For Use With (Application) Category 1 Personal Protective Equipment
Gender Unisex
Size X-Large
Product Type General Purpose Gown
Certifications/Compliance (Europe) EN 1149-5
Construction Made in China
Material Polypropylene
Color Blue
Closure Type Zipper
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Finish Microporous, Anti-Static
Seam Type Sewn
Resistance Waterproof
Certifications/Compliance ISO 9001
Autoclavable Not Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.