Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Medicom™ Hopen™ Single Elastic Bouffant Cap
Click to view available options
Color:
Blue
White
Description
- Simple elastic model provides the user with good support by covering the entirety of their hair
- Possibility of identification by color code
- Intended to be worn by patients and caregivers in the operating room
- Can be used as an object for controlling the environment in HACCP and various other industries
Specifications
Specifications
Material | Polypropylene |
Color | Blue |
Disposable | Single-use |
Sterility | Non-sterile |
For Use With (Equipment) | Operating Room PPE |
Certifications/Compliance | Manufactured under an ISO 9001 Certified Quality System |
Packaging Type | Bag |
Dimensions (L x W) | 44 x 50 cm |
Construction | Elasticated with Welded Ends |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction