Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HOPX Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548205
Description
HOPX Polyclonal Antibody for Western Blot, Flow
HOPX is an atypical homeodomain protein which does not bind DNA and is required to modulate cardiac growth and development. HOPX acts via its interaction with SRF, thereby modulating the expression of SRF-dependent cardiac-specific genes and cardiac development. HOPX prevents SRF-dependent transcription either by inhibiting SRF binding to DNA or by recruiting histone deacetylase (HDAC) proteins that prevent transcription by SRF. HOPX overexpression causes cardiac hypertrophy and may act as a tumor suppressor.
Specifications
HOPX | |
Polyclonal | |
Unconjugated | |
Hopx | |
1110018K11Rik; 1200015P04Rik; 2300002F06Rik; AI848177; AW490897; Cameo; GIIg15b; global ischemia induced protein GIIG15B; global ischemia-induced gene 15B protein; global ischemia-induced protein 15B; Hdop; HOD; homeobox only domain; homeobox-only protein; homeodomain only protein; homeodomain-only protein; Hop; HOP homeobox; Hopx; LAGY; Lung cancer-associated Y protein; mOB1; NECC1; not expressed in choriocarcinoma clone 1; Not expressed in choriocarcinoma protein 1; OB1; Obl; odd homeobox 1; odd homeobox protein 1; SMAP31; Toto | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
84525 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9BPY8 | |
Hopx | |
KLH conjugated synthetic peptide between 39-67 amino acids from the C-terminal region of human HOPX. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction