Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOPX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114744
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114744 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114744 Supplier Invitrogen™ Supplier No. PA5114744
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HOP.

HOPX is an atypical homeodomain protein which does not bind DNA and is required to modulate cardiac growth and development. HOPX acts via its interaction with SRF, thereby modulating the expression of SRF-dependent cardiac-specific genes and cardiac development. HOPX prevents SRF-dependent transcription either by inhibiting SRF binding to DNA or by recruiting histone deacetylase (HDAC) proteins that prevent transcription by SRF. HOPX overexpression causes cardiac hypertrophy and may act as a tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOPX
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Hopx
Gene Accession No. Q8R1H0, Q9BPY8
Gene Alias 1110018K11Rik; 1200015P04Rik; 2300002F06Rik; AI848177; AW490897; Cameo; GIIg15b; global ischemia induced protein GIIG15B; global ischemia-induced gene 15B protein; global ischemia-induced protein 15B; Hdop; HOD; homeobox only domain; homeobox-only protein; homeodomain only protein; homeodomain-only protein; Hop; HOP homeobox; Hopx; LAGY; Lung cancer-associated Y protein; mOB1; NECC1; not expressed in choriocarcinoma clone 1; Not expressed in choriocarcinoma protein 1; OB1; Obl; odd homeobox 1; odd homeobox protein 1; SMAP31; Toto
Gene Symbols Hopx
Host Species Rabbit
Immunogen A synthesized peptide derived from human HOPX(Accession Q9BPY8), corresponding to amino acid residues A40-D73.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 74318, 84525
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.