Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HORIBA ABX Pentra™ 400 Clinical Chemistry Analyzer Accessories: Reagent Cups
Click to view available options
Capacity (Metric):
4 mL
10 mL
15 mL
Description
- Available in 4, 10 and 15mL sizes
Specifications
Specifications
Type | Reagent Cup |
Capacity (Metric) | 15 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction