Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA ABX Pentra™ 400 Clinical Chemistry Analyzer Accessories: Sample Cups

Catalog No. 23600495 Shop All ABX Diagnostics, Inc. Products
Click to view available options
Color:
Blue
Green
White
Yellow

For use with Pentra 400 Clinical Chemistry Analyzer

  • Available in multiple colors

Specifications

Type Sample Cup
For Use With (Equipment) ABX Pentra™ 400
Color Green
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.