Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA ABX Pentra™ 400 Clinical Chemistry Analyzer Calibrators
GSA_VA

Catalog No. 23600441 Shop All ABX Diagnostics, Inc. Products
Click to view available options
Type:
CRP Calibrator
HDL Calibrator
HbAlc WB Calibrator
LDL Calibrator
Multi Calibrator
Quantity:
1 x 8 + 5 x 2 mL
10 x 3 mL
2 x 1 mL
5 x 1 mL
Form:
Lyophilized
This item is not returnable. View return policy
This item is not returnable. View return policy

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage 2°C to 10°C
For Use With (Equipment) ABX Pentra 400 Clinical Chemistry Analyzer
Type Multi Calibrator
Quantity 10 x 3 mL
Form Lyophilized
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.