Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Horizontal Gel Box, 15 cm
SCI_ED

Catalog No. S28806 Shop All Axygen Products
Change view
Click to view available options
Dimensions (L x W):
17.5 x 26.5 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
S28806 17.5 x 26.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S28806 Supplier Axygen™ Supplier No. HGB15/EMD
This item is not returnable. View return policy

The Axygen HGB-15 is large sample capacity system ideal for screening PCR samples or routine DNA fragment analysis and comes standard with two different size UV transparent gel trays providing flexibility and value.

The external gel casting provides efficiency as gel casting and gel running can occur simultaneously. The one piece injection molded tank is durable and leak free while the study electrode cassettes are user changeable. This compact unit is capable of separating bands over a distance of 15 cm. The system only requires 500 mL of buffer making it an economical choice. System includes the novel Axygen contrast enhancer that allows for easy visualization of wells for sample loading and is reversible with white background to visualize running of gels with colored MW markers.
  • Two size gel trays included 15x 10 cm and 15 x 15 cm
  • Axygen Contrast Enhancer
  • User replaceable electrodes
  • Low buffer volume
  • External gel casting
  • Recirculation ports

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE certified
Type Horizontal Gel Box
Volume (Metric) Buffer 500 mL
Dimensions (L x W) 17.5 x 26.5 cm
Width (Metric) 15 cm
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.