Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Horizontal Gel Box, 7 cm
SCI_ED

Catalog No. S28781 Shop All Axygen Products
Change view
Click to view available options
Dimensions (L x W):
7 x 7 cm, 7 x 10 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
S28781 7 x 7 cm, 7 x 10 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S28781 Supplier Axygen™ Supplier No. HGB7/EMD

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

View Alternate Product

This item is not returnable. View return policy

Each unit comes with two sizes of gel trays

  • Feature leak-free external gel casting that allows casting one gel outside of the box while another gel is running inside
  • Easy to replace electrodes that are recessed to reduce potential damage
  • These gel boxes provide years of reliable service
  • Flexibility: Each unit comes with 2 sizes of gel trays to optimize applications
  • External gel casting to gel box allows for running gels while casting the next one
  • Ease of use: the Axygen contrast enhancer makes loading gels easier
  • Rugged injection molding is supported by a 3-year warranty
  • Larger units come with recirculation ports allowing for buffer exchange
TRUSTED_SUSTAINABILITY

Specifications

Dimensions (L x W) 7 x 7 cm, 7 x 10 cm
Length (English) 2.7 in.
Length (Metric) 7 cm
Width (English) 2.7 in., 3.9 in.
Width (Metric) 7 cm, 10 cm
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.