Learn More
Description
- Platinum electrode cassettes can be easily removed for cleaning or replacement
- High quality, heavy duty construction with vast array of accessory options
- Winged gel trays for easy handling. No more sticking fingers into slippery buffer to extract gel tray
- Addition of external gel caster and additional gel tray allows researcher to use time more efficiently
- Tapered baffles allow easy in situ gel formation with or without gel tray
- Run up to 32 samples per gel. Two slots in each gel tray
- Combs are printed with catalog number and thickness for easy identification
- Choice of double-sided adjustable height combs gives researcher more options
- Optional PTFE-coated aluminum combs for low-friction comb removal and accelerated well formation
- Available in four vivid colors; green, yellow, blue or fluorescent pink
- Color-coded horizontal systems are recommended to differentiate exclusive use by specific lab or for specific purpose
Specifications
Specifications
| Certifications/Compliance | CE |
| Color | Pink |
| Description | Compatible Gel Tray: MGT-122 |
| Type | Mini-Gel Kit |
| Gel Dimensions | 8.8 x 5.7 cm |
| Volume (Metric) Buffer | 150 mL |
| Warranty | 3 Years |
| Dimensions (L x W) | Gel Tray I.D.: 8.8 x 5.2 cm (3.4 x 2.04 in.) |
| Dimensions (L x W x H) | 18.5 x 8.5 x 13 cm (7.2 x 3.3 x 5.11 in.) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
