Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HORMAD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558138
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558138 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558138 Supplier Invitrogen™ Supplier No. PA558138
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSESKTRSGK VFQNKMANGN QPVKSSKENR KRSQHESGRI VLHHFDSSSQ ESVPKRRKFS EPK Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 68%.

This gene encodes a HORMA domain-containing protein. HORMA domains are involved in chromatin binding and play a role in cell cycle regulation. The encoded protein may play a role in meiosis, and expression of this gene is a potential marker for cancer. A pseudogene of this gene is located on the long arm of chromosome 6. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HORMAD1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HORMAD1
Gene Accession No. Q86X24
Gene Alias 4921522K05Rik; Cancer/testis antigen 46; CT46; HORM1; HORMA domain containing 1; HORMA domain containing protein; HORMA domain-containing protein 1; HORMAD1; Newborn ovary HORMA protein; Nohma; RGD1564960; testis tissue sperm-binding protein Li 86P
Gene Symbols HORMAD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HORMAD1. Recombinant protein control fragment (Product #RP-94288).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84072
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.