Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HORMAD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | HORMAD1 |
---|---|
Concentration | 0.19 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-9089
![]() ![]() |
Proteintech Group Inc
139171AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes a HORMA domain-containing protein. HORMA domains are involved in chromatin binding and play a role in cell cycle regulation. The encoded protein may play a role in meiosis, and expression of this gene is a potential marker for cancer. A pseudogene of this gene is located on the long arm of chromosome 6. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Specifications
HORMAD1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
D3ZWE7, Q86X24, Q9D5T7 | |
365868, 67981, 84072 | |
HORMAD1 Fusion Protein Ag4914 | |
Primary | |
-20°C | |
HORMAD1 |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Cancer/testis antigen 46, CT46, HORMA domain containing 1, HORMAD1, Newborn ovary HORMA protein, NOHMA, RP11 363I22.1 | |
Hormad1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title