Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HORMAD2 Antibody (OTI3E4), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272416R
Description
HORMAD2 Monoclonal specifically detects HORMAD2 in Human samples. It is validated for Western Blot.Specifications
HORMAD2 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HORMAD2 | |
Full length recombinant protein of human HORMAD2 (NP_689723) produced in HEK293T cells. | |
0.1 mL | |
150280.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI3E4 | |
Western Blot | |
HORMA domain containing 2, HORMA domain-containing protein 2, MGC26710 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction