Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HORMAD2 Antibody (OTI3E4), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272416PCP
Description
HORMAD2 Monoclonal antibody specifically detects HORMAD2 in Human samples. It is validated for Western BlotSpecifications
HORMAD2 | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
150280 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot | |
OTI3E4 | |
Western Blot | |
HORMA domain containing 2, HORMA domain-containing protein 2, MGC26710 | |
Full length human recombinant protein of human HORMAD2(NP_689723) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction