Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HORMAD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139804
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139804 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139804 Supplier Invitrogen™ Supplier No. PA5139804
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: SHFLLFDKEP INVQVGFVST GFHSMKVKVM TEATKVIDLE NNLFRENSTT EIAHQGLDCD EEEECNDHIQ RMNFVCSQQS SECSRKK Highest antigen sequence indentity to the following orthologs: Mouse - 64%, Rat - 50%.

Essential for synapsis surveillance during meiotic prophase via the recruitment of ATR activity. Plays a key role in the male mid-pachytene checkpoint and the female meiotic prophase checkpoint: required for efficient build-up of ATR activity on unsynapsed chromosome regions, a process believed to form the basis of meiotic silencing of unsynapsed chromatin (MSUC) and meiotic prophase quality control in both sexes. Required for the DNA double-strand break-independent, BRCA1-dependent activation of ATR on the sex chromosomes that is essential for normal sex body formation. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen HORMAD2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HORMAD2
Gene Accession No. Q8N7B1
Gene Alias 4930529M09Rik; CT46.2; HORMA domain containing 2; HORMA domain-containing protein 2; Hormad2
Gene Symbols HORMAD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HORMAD2. Recombinant protein control fragment (Product #RP-108847).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150280
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.