Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Hormones: 3,3',5-Triiodo-L-Thyronine
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1000 mg
Description
For research use only. Not intended for therapeutic use.
Specifications
Specifications
| Form | Crystalline powder |
| Product Type | Progesterone |
| Sterility | Sterile |
| pH | 9 |
| Quantity | 100 mg |
| Source | Human |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction