Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Host cell factor C2 Antibody (OTI5H9), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271862B
Description
Host cell factor C2 Monoclonal specifically detects Host cell factor C2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Host cell factor C2 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
HCFC2 | |
Full length recombinant protein of human Host cell factor C2 (NP_037452) produced in HEK293T cells. | |
0.1 mL | |
29915.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI5H9 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
FLJ94012, HCF2, HCF-2C2 factor, host cell factor 2, host cell factor C2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction