Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences Hot start High Fidelity DNA Polymerase 2X Master Mix
SCI_ED

Catalog No. S03144
Change view
Click to view available options
No. of Reactions:
100
500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
S03144 500
S03145 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S03144 Supplier G-Biosciences Supplier No. 7861973
Add to Cart
Add to Cart

Hot start High Fidelity DNA Polymerase 2X Master Mix is used for PCR, Cloning and Diagnostics purposes.

  • G-biosciences hot start high-fidelity DNA Polymerase 2x master mix is ready to use premix which contains hot start high fidelity DNA Polymerase, dNTPs and optimized buffer with stabilizers.
  • The hot start high-fidelity DNA Polymerase possesses 5' - 3' polymerase and 3' - 5' exonuclease (proofreading) activities which has the fidelity, sensitivity and processivity with an error rate ~2.8 x 102-fold lower than Taq DNA polymerase, and significantly lower than other proofreading DNA polymerases.

Specifications

Detection Method qPCR, RT-PCR
GC-Rich PCR Performance High
Reaction Speed Fast or Standard
Product Type DNA Polymerase Master Mix
For Use With (Application) Gene Expression, Genotyping, Microbial Detection, Pathogen Detection, Virus Detection
No. of Reactions 500
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.