Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences Hot Start High Fidelity DNA Polymerase
SCI_ED

Hot start high fidelity DNA Polymerase is used for PCR, cloning, long and high GC content template DNA amplification and diagnostics purposes.

$154.00 - $675.00

Specifications

Detection Method Gel Electrophoresis
Form Liquid
GC-Rich PCR Performance High
PCR Method Standard PCR, Direct PCR, qPCR, Multiplex qPCR, 1-step RT-qPCR
Reaction Speed Fast
View More Specs

Products 2
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
S03146
SCI_ED
View Documents
G-Biosciences
7861971
500
Each for $675.00
Only null left
Add to Cart
 
S03147
SCI_ED
View Documents
G-Biosciences
7861970
100
Each for $154.00
Only null left
Add to Cart
 
Description

Description

  • G-biosciences hot start high-fidelity DNA Polymerase is chemically modified that possesses 5' - 3' polymerase and 3' - 5' exonuclease (proofreading) activities.
  • This Polymerase has the fidelity, sensitivity and processivity with an error rate ~2.8 x 102-fold lower than Taq DNA polymerase, and significantly lower than other proofreading DNA polymerases.
Specifications

Specifications

Gel Electrophoresis
High
Fast
DNA Polymerase
Liquid
Standard PCR, Direct PCR, qPCR, Multiplex qPCR, 1-step RT-qPCR
Biotin, 6-FAM, ROX, SYBR Green, TAMRA
Hot-start PCR, Multiplex PCR, High-fidelity PCR, Site-directed Mutagenesis, Next-Generation Sequencing (NGS)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.