Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Hot Start Mix RTG

Catalog No. 46001004 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
illustra Hot Start Mix RTG
Quantity:
100 Reactions
480 Reactions
96 Reactions
Volume:
200μL
500μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity Volume
46-001-004 illustra Hot Start Mix RTG 480 Reactions 200μL
46-001-003 illustra Hot Start Mix RTG 96 Reactions 200μL
46-001-002 illustra Hot Start Mix RTG 100 Reactions 500μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 46-001-004 Supplier Cytiva Supplier No. 28900654
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Premixed, predispensed, ambient temperature-stable formulation providing robust and reliable performance for demanding PCR applications in which high specificity and high sensitivity are essential to success

  • Primer sequestration effectively blocks greater than 99% of polymerase activity during setup effectively minimizing any nonspecific priming or primer-dimer formation
  • Recombinant activator protein avoids the risk of contamination from a mammalian source associated with hot start antibodies
  • Quick hot start activation preserves sample integrity by avoiding extensive precycling heat treatment
  • The Ready-To-Go manufacturing process locks in performance for excellent reproducibility with less risk of pipetting errors and contamination
  • Long-term ambient-temperature stability simplifies shipping and removes any need for freezer storage

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1.75 units of recombinant PuReTaq DNA Polymerase, dATP, dCTP, dGTP, dTTP, stabilizers, BSA and reaction Buffer. Ambient temperature in pouch with desiccant
Description Preformulated and predispensed, freeze-dried PCR reagents mix including high-quality PuReTaq DNA Polymerase and a hot start activator protein, for increased specificity and reproducibility of PCR amplifications.
Format Tube
Product Type illustra Hot Start Mix RTG
Quantity 480 Reactions
For Use With (Application) Standard PCR
No. of Reactions 480
Volume 200μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.