Learn More
Restek™ Hot Swap Capillary Column Nuts
SDP
Catalog No. 06611208 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Application):
Agilent Compact Ferrules
Agilent Standard 1/16 in. Ferrules
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
06-611-208 Agilent Compact Ferrules
06-611-207 Agilent Standard 1/16 in. Ferrules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-611-208 Supplier Restek™ Supplier No. 22348
For Agilent Ferrules/GCs
  • Reduces the risk of burnt fingers
  • Eliminates downtime waiting for injector parts to cool down
Specifications
Type Hot Swap Capillary Column Nuts
Material Stainless Steel
For Use With (Application) Agilent Compact Ferrules
Quantity 1 Nμt
For Use With (Equipment) Agilent GCs