Learn More
Honeywell™ Howard Leight™ Fusion™ Ear Plugs
Catalog No. 19100152
Click to view available options
Size:
Regular
Small
Catalog No. 19-100-152
Supplier: Honeywell™ FUS30SHP
Fusion™ advanced manufacturing technology delivers a combination of comfort and easy handling. Detachable nylon cord system can be used in a variety of applications.
  • Firm core makes insertion and removal easy
  • Constructed of soft outer flanges thermally fused to a firm inner core
  • Patented quadruple SoftFlange™ design offers excellent attenuation
  • FlexiFirm™ core makes it simple to achieve a proper fit that stays in place
  • Fabric cord detaches easily to alternate between corded and uncorded applications
TRUSTED_SUSTAINABILITY
Specifications
ANSI S3.1974 Yes
Cord Color Blue, yellow
Cord Material Nylon
CSA Z94.2 Class AL
Size Small
For Use With (Application) Facility Maintenance, Grinding, Electrical, Welding, Demolition, Painting, Blasting, Assembly, Cleaning
High Visibility No
Packaging Type HearPack case
Pairs per Package 100 Pairs/Pk.
Plug Material TPE
Plug Color Blue, yellow
Noise Reduction Rating (NRR) 27 dB
Corded Corded
Plug Shape Flanged
Disposable Reusable
Metal Detectable No
Includes Pocket-size case
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.