Learn More
Honeywell™ Howard Leight™ Quiet Bands HYG Replacement Pods
Catalog No. 19038481 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Model:
QB100HYG
QB200HYG
Noise Reduction Rating (NRR):
25 dB
27 dB
Plug Shape:
Bell
Cone
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Noise Reduction Rating (NRR) Plug Shape
19-038-481 QB200HYG 25 dB Cone
19-038-480 QB100HYG 27 dB Bell
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-038-481 Supplier Honeywell™ Supplier No. QB200HYG
This item is not returnable. View return policy
Replacement pods for the Howard Leight™ Quiet Bands HYG models QB1HYG, QB2HYG and QB3HYG.
  • Replacement pods for the Howard Leight™ quiet bands HYG models
  • NRR 27 dB QB100HYG, NRR 25 dB QB200HYG, NRR dB QB300HYG
  • Packs of 50 pairs
  • One size fits all
TRUSTED_SUSTAINABILITY
Specifications
ANSI S3.1974 Yes
Size Universal
For Use With (Application) Replacement Tips for QB2HYG Banded Ear Plugs
Model QB200HYG
Packaging Type Poly bag
Pairs per Package 50 Pairs/Pk.
Plug Material Foam
Plug Color Orange, Blue
Noise Reduction Rating (NRR) 25 dB
Plug Shape Cone
Metal Detectable No