Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXA1/HOXB1/HOXD1 Monoclonal Antibody (2F12)
GREENER_CHOICE

Catalog No. PIMA519110
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA519110 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA519110 Supplier Invitrogen™ Supplier No. MA519110
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: LGQSEGDGGY FHPSSYGAQL GGLSDGYGAG GAGPGPYPPQ HPPYGNEQTA SFAPAYADLL SEDKETPCPS EPNTPTARTF DWMKVKRNPP KTAKVSEPGL GSPSGLRTNF.

This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXB genes located in a cluster on chromosome 17.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXA1/HOXB1/HOXD1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2F12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene HOXB1
Gene Accession No. P14653, P49639, Q9GZZ0
Gene Alias BSAS; early retinoic acid 1; ERA1; HCFP3; homeo box 4G; homeo box A1; homeo box B1; homeo box D1; homeobox 1F; homeobox A1; homeobox B1; homeobox D1; homeobox protein Hox-1.6; homeobox protein Hox-1F; Homeobox protein Hox-2.9; homeobox protein Hox-2I; Homeobox protein Hox-4.9; homeobox protein Hox-A1; Homeobox protein Hox-B1; homeobox protein Hox-D1; Homeobox protein Hox-GG; homeoboxless protein ERA-1-399; homeotic protein ERA-1-993; Hox 1.6-like protein; HOX A1 homeodomain protein; HOX1; Hox-1.6; HOX1F; HOX2; Hox-2.9; HOX2I; HOX4; Hox-4.7; Hox-4.9; HOX4G; HOXA1; Hoxb1; Hoxb-1; Hoxd1; Hoxd-1; lab-like protein; MGC45232
Gene Symbols HOXA1, HOXB1, HOXD1
Host Species Mouse
Immunogen HOXB1 (NP_002135.2, 101 a.a. ~ 210 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3198, 3211, 3231
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.