Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXB1 Polyclonal Antibody
HOXA1/HOXB1/HOXD1 Polyclonal Antibody
Supplier: Thermo Scientific PA5103889
Description
HOXA1/HOXB1/HOXD1 Polyclonal Antibody for Western Blot, IHC (P)

Specifications
HOXB1 | |
Polyclonal | |
Unconjugated | |
Hoxb1 | |
HCFP3; homeo box B1; homeobox B1; homeobox protein Hox-2.9; Homeobox protein Hox-2I; homeobox protein Hox-B1; HOX2; Hox-2.9; Hox-29; HOX2I; Hoxb1; Hoxb-1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
15407, 3211 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P14653, P17919 | |
Hoxb1 | |
A synthesized peptide derived from human HOXB1(Accession P14653), corresponding to amino acid residues S78-G128. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction