Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXA1/HOXB1/HOXD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538578
Description
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. HOXA1 is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. The encoded protein may be involved in the placement of hindbrain segments in the proper location along the anterior-posterior axis during development. Two transcript variants encoding two different isoforms have been found for this gene, with only one of the isoforms containing the homeodomain region.
Specifications
HOXA1/HOXB1/HOXD1 | |
Polyclonal | |
Unconjugated | |
HOXA1 | |
HOXA1; homeobox A1; BSAS; HOX1; HOX1F; MGC45232; HOX A1 homeodomain protein; Hox 1.6-like protein; homeo box A1; homeobox 1F; homeobox protein HOX-A1; lab-like protein, Homeobox protein Hox-B1, Homeobox protein Hox-2I, HOX2I | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15394, 15407, 15429, 3198, 3211, 3231 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P14653, P49639, Q9GZZ0, P09022, P17919, Q01822 | |
HOXA1, HOXB1, HOXD1 | |
A synthetic peptide derived from the internal region of human HOXA1/B1/D1 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction