Learn More
Invitrogen™ HOXA1 Polyclonal Antibody

Description
This antibody detects endogenous protein at a molecular weight of 37 kDa. Purity is >95% by SDS-PAGE.
Specifications
Specifications
| Antigen | HOXA1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | HOXA1 |
| Gene Accession No. | P49639 |
| Gene Alias | BSAS; early retinoic acid 1; ERA1; homeo box A1; homeobox 1F; homeobox A1; homeobox protein Hox-1.6; homeobox protein Hox-1F; Homeobox protein Hox-A1; homeoboxless protein ERA-1-399; homeotic protein ERA-1-993; Hox 1.6-like protein; HOX A1 homeodomain protein; HOX1; Hox-1.6; HOX1F; HOXA 1; HOXA1; HOXA-1; lab-like protein; MGC45232; NEWGENE_2812 |
| Gene Symbols | HOXA1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.