Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597835
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597835 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597835 Supplier Invitrogen™ Supplier No. PA597835
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HOXA1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. The encoded protein may be involved in the placement of hindbrain segments in the proper location along the anterior-posterior axis during development. Two transcript variants encoding two different isoforms have been found for this gene, with only one of the isoforms containing the homeodomain region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXA1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HOXA1
Gene Accession No. P49639
Gene Alias BSAS; early retinoic acid 1; ERA1; homeo box A1; homeobox 1F; homeobox A1; homeobox protein Hox-1.6; homeobox protein Hox-1F; Homeobox protein Hox-A1; homeoboxless protein ERA-1-399; homeotic protein ERA-1-993; Hox 1.6-like protein; HOX A1 homeodomain protein; HOX1; Hox-1.6; HOX1F; HOXA 1; HOXA1; HOXA-1; lab-like protein; MGC45232; NEWGENE_2812
Gene Symbols HOXA1
Host Species Rabbit
Immunogen Recombinant Human Homeobox protein Hox-A1 protein (75-205AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3198
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.