Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXA10 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA523781
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523781 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523781 Supplier Thermo Scientific Supplier No. PA523781

Rabbit Polyclonal Antibody

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factorm HOXA10, that may regulate gene expression, morphogenesis, and differentiation. More specifically, HOXA10 may function in fertility, embryo viability, and regulation of hematopoietic lineage commitment. Alternatively spliced transcript variants have been described. Read-through transcription also exists between this gene and the downstream homeobox A9 gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXA10
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene HOXA10
Gene Accession No. P31260
Gene Alias homeo box A10; homeobox A10; homeobox protein 1H; homeobox protein Hox-1.8; Homeobox protein Hox-1H; homeobox protein HOXA10; homeobox protein Hox-A10; HOX1; HOX1.8; Hox-1.8; Hox-18; HOX1H; HOXA10; Hoxa-10; PL
Gene Symbols HOXA10
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 244-271 amino acids from the central region of human HOXA10
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3206
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype Ig
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.