Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXA10 Polyclonal Antibody, PE, Bioss
SDP

Catalog No. 502281907
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-1907 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-228-1907 Supplier Bioss Supplier No. BS2502RPE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HOXA10 Polyclonal antibody specifically detects HOXA10 in Human, Mouse, Rat samples. It is validated for Flow Cytometry

Specifications

Antigen HOXA10
Applications Flow Cytometry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate PE
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene HOXA10
Gene Accession No. P31260, P31310
Gene Alias homeo box A10; homeobox A10; homeobox protein 1H; homeobox protein Hox-1.8; Homeobox protein Hox-1H; homeobox protein HOXA10; homeobox protein Hox-A10; HOX1; HOX1.8; Hox-1.8; Hox-18; HOX1H; HOXA10; Hoxa-10; PL
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HOXA10
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103692129, 15395, 3206
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.