Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXA11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570380
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570380 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570380 Supplier Invitrogen™ Supplier No. PA570380
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 85%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Sheep: 85%; Zebrafish: 93%.

HOX11, also called homeobox11 or Tcl-3 is located at the 10q24 T-Cell translocation breakpoint region. It encodes a homeobox-domain containing protein, HOX11, containing a glycine and proline-rich amino terminus. HOX11 is required for maintenance of the developing spleen and cell survival. The HOX11protein binds to a specific DNA sequence and localizes to the cell nucleus. It transactivates transcription of a reporter gene linked to a cis-regulatory element, acting as a positive transcription activator. The HOX11 gene is highly expressed in T-ALLs as a result of a t (7:10) (q35;q24) or t (10:14) (q24;qll) translocation. The homeobox gene is deregulated upon translocation into the T-cell receptor locus and activates a specific subset of tumor-associated target genes. HOX 11 gene translocation has been attributed to T-cell leukemia and lymphoid neoplasias.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXA11
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HOXA11
Gene Accession No. 0
Gene Alias homeo box 1I; homeo box A11; homeobox A11; homeobox protein Hox-1.9; Homeobox protein Hox-1I; homeobox protein HOXA11; homeobox protein Hox-A11; HOX1; Hox-1.9; HOX11; HOX1I; Hoxa11; Hoxa-11; NEWGENE_1564605; RUSAT1
Gene Symbols HOXA11
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human HOXA11.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103692131
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.