Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXA13 Antibody (SR1709), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | HOXA13 |
---|---|
Clone | SR1709 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
HOXA13 Monoclonal antibody specifically detects HOXA13 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
HOXA13 | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
homeo box 1J, homeo box A13, homeobox A13, Homeobox protein Hox-1J, homeobox protein HOXA13, HOX1, HOX1Jhomeobox protein Hox-A13, transcription factor HOXA13 | |
A synthesized peptide derived from human HOXA13 (Uniprot #: P31271) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1709 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Biology | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
3209 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title