Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HOXA13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5139894
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LGLPMESYQP WALPNGWNGQ MYCPKEQAQP PHLWKSTLPD VVSHPSDASS Y.
HoxA13 and HoxD13 also bind to other BMP and TGF-beta/Activin-regulated Smad proteins including Smad1 and Smad2, but not Smad4. In humans and mice, loss of HOXA13 function causes defects in the growth and patterning of the digits and interdigital tissues. Analysis of HoxA13 expression reveals a pattern of localization overlapping with sites of reduced Bmp2 and Bmp7 expression in HoxA13 mutant limbs. HoxA13 regulates Bmp2 and Bmp7 expression, providing a link between HoxA13, its target genes and the specific developmental processes affected by loss of HoxA13 function.
Specifications
HOXA13 | |
Polyclonal | |
Unconjugated | |
Hoxa13 | |
Hd; homeo box 1J; homeo box A13; homeobox A13; homeobox protein Hox-1.10; Homeobox protein Hox-1J; homeobox protein HOXA13; homeobox protein Hox-A13; HOX1; Hox-1.10; HOX1J; HOXA13; hypodactyly; RGD1562483; transcription factor HOXA13 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3209 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P31271 | |
Hoxa13 | |
Recombinant protein corresponding to Human HOXA13. Recombinant protein control fragment (Product #RP-109174). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction