Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXA13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540459
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540459 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540459 Supplier Invitrogen™ Supplier No. PA540459
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: AAAAANQCRN LMAHPAPLAP GAASAYSSAP GEAPPSAAAA AAAAAAAAAA Sequence homology: Dog: 83%; Human: 100%; Mouse: 92%.

HoxA13 and HoxD13 also bind to other BMP and TGF-beta/Activin-regulated Smad proteins including Smad1 and Smad2, but not Smad4. In humans and mice, loss of HOXA13 function causes defects in the growth and patterning of the digits and interdigital tissues. Analysis of HoxA13 expression reveals a pattern of localization overlapping with sites of reduced Bmp2 and Bmp7 expression in HoxA13 mutant limbs. HoxA13 regulates Bmp2 and Bmp7 expression, providing a link between HoxA13, its target genes and the specific developmental processes affected by loss of HoxA13 function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXA13
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Hoxa13
Gene Accession No. P31271
Gene Alias Hd; homeo box 1J; homeo box A13; homeobox A13; homeobox protein Hox-1.10; Homeobox protein Hox-1J; homeobox protein HOXA13; homeobox protein Hox-A13; HOX1; Hox-1.10; HOX1J; HOXA13; hypodactyly; RGD1562483; transcription factor HOXA13
Gene Symbols Hoxa13
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human HOXA13 (aa 80-129).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3209
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.