Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXA1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572624
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572624 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572624 Supplier Invitrogen™ Supplier No. PA572624
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HOXA1A Polyclonal Antibody for Western Blot

Sequence-specific transcription factor. Part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXA1A
Applications Western Blot
Classification Polyclonal
Concentration 0.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene hoxa1a
Gene Accession No. Q98SI1
Gene Alias homeo box A1a; homeobox a1a; homeobox protein Hox-A1a; hoxa1; Hox-a1; hoxa1a; Hoxa1a protein; hoxa3a; id:ibd3526; z-4
Gene Symbols hoxa1a
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 192-219 amino acids from the Central region of DANRE hoxa1a.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58051
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.