Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HOXA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109830
Description
Immunogen sequence: NQNSEGKCKS LEDSEKVEED EEEKTLFEQA LSVSGALLER EGYTFQQNAL SQQQAPNGHN GDSQSFPVSP LTSNEKNLKH FQHQSPTVPN CLSTMGQNCG AGLNNDSPEA LEVPSLQDFS VFSTDSCLQL SDAVSPSLPG SLDSPVDISA DSLDFFTDTL TTIDLQHLNY.
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.
Specifications
HOXA2 | |
Polyclonal | |
Unconjugated | |
HOXA2 | |
AI324701; homeo box A2; homeobox A2; homeobox protein Hox-1.11; Homeobox protein Hox-1K; homeobox protein Hox-A2; Hox1.11; Hox-1.11; Hox111; Hox-111; HOX1K; HOXA2; Hoxa-2; MCOHI; NEWGENE_2813 | |
Rabbit | |
Affinity chromatography | |
RUO | |
15399, 3199 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43364, P31245 | |
HOXA2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 207-376 of human HOXA2 (NP_006726.1). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction