Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXA3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA526887

Catalog No. PIPA526887


Only null left
Add to Cart

Description

Description

This antibody is predicted to react with bovine, chicken, mouse and zebrafish based on sequence homology.

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HOXA3
Polyclonal
Unconjugated
Hoxa3
homeo box 1E; homeo box A3; homeobox A2; homeobox A3; homeobox protein Hox-1.5; Homeobox protein Hox-1E; homeobox protein Hox-A3; Homeobox protein MO-10; HOX1; Hox-1.5; Hox-1.5-like protein; Hox-15; HOX1E; HOXA3; Hoxa-3; Mo-10; NEWGENE_1561431
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
3200
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
O43365
Hoxa3
KLH conjugated synthetic peptide between 413-440 amino acids from the C-terminal region of human HOXA3.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.