Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOXA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568810
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568810 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568810 Supplier Invitrogen™ Supplier No. PA568810
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 86%; Guinea Pig: 83%; Horse: 92%; Human: 93%; Mouse: 100%; Pig: 93%; Rabbit: 85%; Rat: 93%.

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXA3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Hoxa3
Gene Accession No. 0
Gene Alias homeo box 1E; homeo box A3; homeobox A2; homeobox A3; homeobox protein Hox-1.5; Homeobox protein Hox-1E; homeobox protein Hox-A3; Homeobox protein MO-10; HOX1; Hox-1.5; Hox-1.5-like protein; Hox-15; HOX1E; HOXA3; Hoxa-3; Mo-10; NEWGENE_1561431
Gene Symbols Hoxa3
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of rat HOXA3.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103690130
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.